missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4'-[N-Ethyl-N-(2-methylphenyl)sulfamoyl]acetanilide, 97%, Alfa Aesar™

Chemical Identifiers

CAS 333430-43-6
Molecular Formula C17H20N2O3S
Molecular Weight (g/mol) 332.418
MDL Number MFCD01213966
InChI Key NGMYZQIEURPXLI-UHFFFAOYSA-N
Synonym n-4-ethyl 2-methylphenyl sulfamoyl phenyl acetamide, n-4-ethyl-o-tolyl-sulfamoyl-phenyl-acetamide, 4'-n-ethyl-n-2-methylphenyl sulfamoyl acetanilide, n-4-ethyl-2-methylphenyl sulfamoyl phenyl acetamide, n-4-ethyl-2-methylanilino sulfonyl phenyl acetamide, n-4-ethyl 2-methylphenyl amino sulfonyl phenyl acetamide
PubChem CID 743585
IUPAC Name N-[4-[ethyl-(2-methylphenyl)sulfamoyl]phenyl]acetamide
SMILES CCN(C1=CC=CC=C1C)S(=O)(=O)C2=CC=C(C=C2)NC(=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60904MD View Documents Alfa Aesar
H60904MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6090403 View Documents Alfa Aesar
H6090403
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

333430-43-6
332.418
NGMYZQIEURPXLI-UHFFFAOYSA-N
743585
CCN(C1=CC=CC=C1C)S(=O)(=O)C2=CC=C(C=C2)NC(=O)C
C17H20N2O3S
MFCD01213966
n-4-ethyl 2-methylphenyl sulfamoyl phenyl acetamide, n-4-ethyl-o-tolyl-sulfamoyl-phenyl-acetamide, 4'-n-ethyl-n-2-methylphenyl sulfamoyl acetanilide, n-4-ethyl-2-methylphenyl sulfamoyl phenyl acetamide, n-4-ethyl-2-methylanilino sulfonyl phenyl acetamide, n-4-ethyl 2-methylphenyl amino sulfonyl phenyl acetamide
N-[4-[ethyl-(2-methylphenyl)sulfamoyl]phenyl]acetamide

Specifications

250mg
C17H20N2O3S
n-4-ethyl 2-methylphenyl sulfamoyl phenyl acetamide, n-4-ethyl-o-tolyl-sulfamoyl-phenyl-acetamide, 4'-n-ethyl-n-2-methylphenyl sulfamoyl acetanilide, n-4-ethyl-2-methylphenyl sulfamoyl phenyl acetamide, n-4-ethyl-2-methylanilino sulfonyl phenyl acetamide, n-4-ethyl 2-methylphenyl amino sulfonyl phenyl acetamide
CCN(C1=CC=CC=C1C)S(=O)(=O)C2=CC=C(C=C2)NC(=O)C
332.418
332.42
4'-[N-Ethyl-N-(2-methylphenyl)sulfamoyl]acetanilide
333430-43-6
MFCD01213966
NGMYZQIEURPXLI-UHFFFAOYSA-N
N-[4-[ethyl-(2-methylphenyl)sulfamoyl]phenyl]acetamide
743585
97%