missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-n-Dodecyloxybenzaldehyde, 98%
CAS: 24083-19-0 | C19H30O2 | 290.447 g/mol
$106.63 - $312.47
Chemical Identifiers
| CAS | 24083-19-0 |
|---|---|
| Molecular Formula | C19H30O2 |
| Molecular Weight (g/mol) | 290.447 |
| MDL Number | MFCD00043523 |
| InChI Key | ZBEGLEYBWGNZJA-UHFFFAOYSA-N |
| Synonym | p-dodecyloxybenzaldehyde, 4-dodecyloxy benzaldehyde, 4-dodecyloxybenzaldehyde, 4-n-dodecyloxybenzaldehyde, p-n-dodecoxy benzaldehyde, p-dodecyloxy benzaldehyde, 4-dodecyloxy-benzaldehyde, p-lauryloxy benzaldehyde, 1-4-formylphenoxy dodecane |
| PubChem CID | 141069 |
| IUPAC Name | 4-dodecoxybenzaldehyde |
| SMILES | CCCCCCCCCCCCOC1=CC=C(C=C1)C=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0888903
|
Thermo Scientific Chemicals
L0888903 |
1 g |
Each for $106.63
|
|
|||||
|
AAL0888906
|
Thermo Scientific Chemicals
L0888906 |
5 g |
Each for $312.47
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 24083-19-0 | |
| 290.447 | |
| ZBEGLEYBWGNZJA-UHFFFAOYSA-N | |
| 141069 | |
| CCCCCCCCCCCCOC1=CC=C(C=C1)C=O |
| C19H30O2 | |
| MFCD00043523 | |
| p-dodecyloxybenzaldehyde, 4-dodecyloxy benzaldehyde, 4-dodecyloxybenzaldehyde, 4-n-dodecyloxybenzaldehyde, p-n-dodecoxy benzaldehyde, p-dodecyloxy benzaldehyde, 4-dodecyloxy-benzaldehyde, p-lauryloxy benzaldehyde, 1-4-formylphenoxy dodecane | |
| 4-dodecoxybenzaldehyde |
Specifications
| 24083-19-0 | |
| 165°C (0.005 mmHg) | |
| MFCD00043523 | |
| 2535031 | |
| p-dodecyloxybenzaldehyde, 4-dodecyloxy benzaldehyde, 4-dodecyloxybenzaldehyde, 4-n-dodecyloxybenzaldehyde, p-n-dodecoxy benzaldehyde, p-dodecyloxy benzaldehyde, 4-dodecyloxy-benzaldehyde, p-lauryloxy benzaldehyde, 1-4-formylphenoxy dodecane | |
| CCCCCCCCCCCCOC1=CC=C(C=C1)C=O | |
| 290.447 | |
| 290.45 | |
| 4-n-Dodecyloxybenzaldehyde |
| 26°C to 29°C | |
| C19H30O2 | |
| 1 g | |
| Air Sensitive | |
| ZBEGLEYBWGNZJA-UHFFFAOYSA-N | |
| 4-dodecoxybenzaldehyde | |
| 141069 | |
| 98% |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only