Learn More
4-n-Decyloxybenzoic acid, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1470314
Description
4-n-Decyloxybenzoic acid, is used in the syntheis of liquid crystals. It can also be used to react with 3-bromo-propene to produce 5-allyloxy-2-phenyl-[1,3]dioxane. This reaction will need reagent tetra-n-butylammonium bromide and NaH, and the menstruum dimethylformamide. The reaction time is 2 hour with temperature of 20°C,
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-n-Decyloxybenzoic acid, is used in the syntheis of liquid crystals. It can also be used to react with 3-bromo-propene to produce 5-allyloxy-2-phenyl-[1,3]dioxane. This reaction will need reagent tetra-n-butylammonium bromide and NaH, and the menstruum dimethylformamide. The reaction time is 2 hour with temperature of 20°C,
Solubility
Soluble in water.
Notes
m.p. 97°(S), 122°(N), 142°(I)
Specifications
4-n-Decyloxybenzoic acid | |
97°C (S) | |
MFCD00020360 | |
2113730 | |
Soluble in water. | |
CCCCCCCCCCOC1=CC=C(C=C1)C(=O)O | |
278.392 | |
278.39 |
5519-23-3 | |
C17H26O3 | |
25 g | |
4-decyloxy benzoic acid, p-decyloxybenzoic acid, 4-n-decyloxybenzoic acid, 4-decyloxybenzoic acid, benzoic acid,4-decyloxy, benzoic acid, 4-decyloxy, pubchem2671, p-n-decyloxybenzoic acid, 4-decyloxy benzoicacid, para-decyloxy benzoic acid | |
NZNICZRIRMGOFG-UHFFFAOYSA-N | |
4-decoxybenzoic acid | |
138527 | |
96% |
Chemical Identifiers
5519-23-3 | |
278.392 | |
NZNICZRIRMGOFG-UHFFFAOYSA-N | |
4-decoxybenzoic acid |
C17H26O3 | |
MFCD00020360 | |
138527 | |
CCCCCCCCCCOC1=CC=C(C=C1)C(=O)O |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only