Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Decylbiphenyl, 98%

$131.61 - $504.88

Chemical Identifiers

Chemical Name or Material 4-n-Decylbiphenyl
CAS 93972-02-2
Molecular Formula C22H30
Formula Weight 294.48
MDL Number MFCD00102117
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1668503

 
alfa aesar™
A1668503
1g Each for $131.61

AAA1668506

 
alfa aesar™
A1668506
5g Each for $504.88
Description & Specifications

Specifications

Formula Weight 294.48
Chemical Name or Material 4-n-Decylbiphenyl
Assay 98%
Molecular Formula C22H30
Melting Point 52° to 54°C
MDL Number MFCD00102117
CAS 93972-02-2
Boiling Point 204° to 210°C (1.5mmHg)
Quantity 1g

Applications
PDLC monomer. As chemical reagent.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.