missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(n-Butylsulfamoyl)benzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H5321803

 View more versions of this product

Catalog No. AAH5321803



Specifications

Specifications

4-(n-Butylsulfamoyl)benzeneboronic acid
1g
C10H16BNO4S
n-butyl 4-boronobenzenesulfonamide, 4-n-butylsulfamoyl phenyl boronic acid, 4-n-butylsulphonamido benzeneboronic acid, 4-butylsulfamoyl phenylboronic acid, acmc-209q2v, 4-butylsulfamoyl phenyl boronic acid, n-n-butyl 4-boronobenzenesulfonamide, 4-n-butylsulfamoyl phenylboronic acid, 4-n-n-butylsulfamoylphenylboronic acid, 4-n-butylsulfamoyl benzeneboronic acid
B(C1=CC=C(C=C1)S(=O)(=O)NCCCC)(O)O
257.111
257.11
124°C to 126°C
850589-32-1
MFCD07363753
BEBIGVQNUYZEPK-UHFFFAOYSA-N
[4-(butylsulfamoyl)phenyl]boronic acid
44119412
97%

Chemical Identifiers

850589-32-1
257.111
BEBIGVQNUYZEPK-UHFFFAOYSA-N
44119412
B(C1=CC=C(C=C1)S(=O)(=O)NCCCC)(O)O
C10H16BNO4S
MFCD07363753
n-butyl 4-boronobenzenesulfonamide, 4-n-butylsulfamoyl phenyl boronic acid, 4-n-butylsulphonamido benzeneboronic acid, 4-butylsulfamoyl phenylboronic acid, acmc-209q2v, 4-butylsulfamoyl phenyl boronic acid, n-n-butyl 4-boronobenzenesulfonamide, 4-n-butylsulfamoyl phenylboronic acid, 4-n-n-butylsulfamoylphenylboronic acid, 4-n-butylsulfamoyl benzeneboronic acid
[4-(butylsulfamoyl)phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents