Please login to your online account to display your discounted pricing

4-n-Butylphenylboronic acid, 95%, Acros Organics


Quantity 1g

Manufacturer: thermo fisher scientific  AC393870010

 View more versions of this product

Catalog No. AC393870010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 178.04
Quantity 1g
Assay Percent Range 95%
Chemical Name or Material 4-n-Butylphenylboronic acid
Molecular Formula C10H15BO2
CAS 145240-28-4

Specifications

Chemical Name or Material 4-n-Butylphenylboronic acid
Assay Percent Range 95%
Molecular Formula C10H15BO2
Formula Weight 178.04