Please login to your online account to display your discounted pricing

4-n-Butylphenylboronic acid, 95%, Acros Organics

Chemical Identifiers

Chemical Name or Material 4-n-Butylphenylboronic acid
Assay Percent Range 95%
Molecular Formula C10H15BO2
Formula Weight 178.04
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC393870010

 
thermo fisher scientific
AC393870010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC393870050

 
thermo fisher scientific
AC393870050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 178.04
Quantity 1g
Chemical Name or Material 4-n-Butylphenylboronic acid
Assay Percent Range 95%
Molecular Formula C10H15BO2
CAS 145240-28-4