missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Butylphenyl isocyanate, 98%

Manufacturer:  Alfa Aesar™ L0995714

 View more versions of this product

Catalog No. AAL0995714



Specifications

Specifications

4-n-Butylphenyl isocyanate
107°C to 108°C (3mmHg)
69342-47-8
MFCD00013882
390859
LJJRXPXDTAUVQU-UHFFFAOYSA-N
1-butyl-4-isocyanatobenzene
2796313
98%
108°C (226°F)
25g
C11H13NO
UN2206
4-butylphenyl isocyanate, 4-n-butylphenyl isocyanate, benzene, 1-butyl-4-isocyanato, benzene,1-butyl-4-isocyanato, 4-butylbenzenisocyanate, 4-butylphenylisocyanate, acmc-209o7f, 1-butyl-4-isocyanato-benzene, 4-n-butyl phenyl isocyanate, 4-butylphenyl isocyanate
CCCCC1=CC=C(C=C1)N=C=O
175.231
175.23

Chemical Identifiers

69342-47-8
175.231
LJJRXPXDTAUVQU-UHFFFAOYSA-N
2796313
CCCCC1=CC=C(C=C1)N=C=O
C11H13NO
MFCD00013882
4-butylphenyl isocyanate, 4-n-butylphenyl isocyanate, benzene, 1-butyl-4-isocyanato, benzene,1-butyl-4-isocyanato, 4-butylbenzenisocyanate, 4-butylphenylisocyanate, acmc-209o7f, 1-butyl-4-isocyanato-benzene, 4-n-butyl phenyl isocyanate, 4-butylphenyl isocyanate
1-butyl-4-isocyanatobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.

Recommended Storage : 2° to 8°C

Documents