Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Butylbiphenyl, 97%

$56.05 - $195.88

Chemical Identifiers

Chemical Name or Material 4-n-Butylbiphenyl
CAS 37909-95-8
Molecular Formula C16H18
Formula Weight 210.32
MDL Number MFCD00100665
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1746503

  SDS
alfa aesar™
A1746503
1g Each for $56.05

AAA1746506

  SDS
alfa aesar™
A1746506
5g Each for $195.88
Description & Specifications

Specifications

Formula Weight 210.32
Quantity 1g
Chemical Name or Material 4-n-Butylbiphenyl
Assay 97%
CAS 37909-95-8
Molecular Formula C16H18
Flash Point >110°C (230°F)
Melting Point 6° to 8°C
MDL Number MFCD00100665
Boiling Point 318° to 319°C