missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Butylbenzoyl chloride, 98%

Manufacturer:  Alfa Aesar™ A1508206

 View more versions of this product

Catalog No. AAA1508206

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Applications
4-Butylbenzoyl chloride was used in the synthesis of 2,5-bis[(4-butylbenzoyl)oxy]styrene (BBOS, monomer).

Solubility
Reacts with water.

Notes
Moisture Sensitive, store away from moisture. Store in cool, dry place in tightly closed container. Store away from oxidizing agent. Store under inert gas.
Specifications

Specifications

4-n-Butylbenzoyl chloride
145°C to 147°C (11mmHg)
28788-62-7
MFCD00000699
4-n-butylbenzoyl chloride, benzoyl chloride, 4-butyl, p-butylbenzoyl chloride, 4-n-butyl-benzoyl chloride, 4-butylbenzoylchloride, 4-n-butylbenzoylchloride, p-n-butyl benzoylchloride, p-n-butylbenzoyl chloride, 4-1-butylbenzoyl chloride, acmc-209h4o
CCCCC1=CC=C(C=C1)C(=O)Cl
196.674
196.68
>110°C (230°F)
5g
C11H13ClO
UN3265
OUOWCSJYDCPVDM-UHFFFAOYSA-N
4-butylbenzoyl chloride
120049
98%

Chemical Identifiers

28788-62-7
196.674
OUOWCSJYDCPVDM-UHFFFAOYSA-N
120049
CCCCC1=CC=C(C=C1)C(=O)Cl
C11H13ClO
MFCD00000699
4-n-butylbenzoyl chloride, benzoyl chloride, 4-butyl, p-butylbenzoyl chloride, 4-n-butyl-benzoyl chloride, 4-butylbenzoylchloride, 4-n-butylbenzoylchloride, p-n-butyl benzoylchloride, p-n-butylbenzoyl chloride, 4-1-butylbenzoyl chloride, acmc-209h4o
4-butylbenzoyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

TSCA : TSCA

Documents