Learn More
Alfa Aesar™ 4-n-Butylbenzoyl chloride, 98%
Manufacturer: Alfa Aesar™ A1508222
Description
Applications4-Butylbenzoyl chloride was used in the synthesis of 2,5-bis[(4-butylbenzoyl)oxy]styrene (BBOS, monomer).
Solubility
Reacts with water.
Notes
Moisture Sensitive, store away from moisture. Store in cool, dry place in tightly closed container. Store away from oxidizing agent. Store under inert gas.
Specifications
4-n-Butylbenzoyl chloride | |
145°C to 147°C (11mmHg) | |
28788-62-7 | |
MFCD00000699 | |
4-n-butylbenzoyl chloride, benzoyl chloride, 4-butyl, p-butylbenzoyl chloride, 4-n-butyl-benzoyl chloride, 4-butylbenzoylchloride, 4-n-butylbenzoylchloride, p-n-butyl benzoylchloride, p-n-butylbenzoyl chloride, 4-1-butylbenzoyl chloride, acmc-209h4o | |
CCCCC1=CC=C(C=C1)C(=O)Cl | |
196.674 | |
196.68 |
>110°C (230°F) | |
100g | |
C11H13ClO | |
UN3265 | |
OUOWCSJYDCPVDM-UHFFFAOYSA-N | |
4-butylbenzoyl chloride | |
120049 | |
98% |
Chemical Identifiers
28788-62-7 | |
196.674 | |
OUOWCSJYDCPVDM-UHFFFAOYSA-N | |
120049 | |
CCCCC1=CC=C(C=C1)C(=O)Cl |
C11H13ClO | |
MFCD00000699 | |
4-n-butylbenzoyl chloride, benzoyl chloride, 4-butyl, p-butylbenzoyl chloride, 4-n-butyl-benzoyl chloride, 4-butylbenzoylchloride, 4-n-butylbenzoylchloride, p-n-butyl benzoylchloride, p-n-butylbenzoyl chloride, 4-1-butylbenzoyl chloride, acmc-209h4o | |
4-butylbenzoyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA : TSCA