Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Butylbenzonitrile, 98%

$101.70 - $1,315.41

Chemical Identifiers

Chemical Name or Material 4-n-Butylbenzonitrile
CAS 20651-73-4
Molecular Formula C11H13N
Formula Weight 159.23
MDL Number MFCD00052345
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1917406

  SDS
alfa aesar™
A1917406
5g Each for $101.70

AAA1917414

  SDS
alfa aesar™
A1917414
25g Each for $382.72

AAA1917422

  SDS
alfa aesar™
A1917422
100g Each for $1,315.41
Description & Specifications

Specifications

Formula Weight 159.23
Chemical Name or Material 4-n-Butylbenzonitrile
Assay 98%
Molecular Formula C11H13N
MDL Number MFCD00052345
CAS 20651-73-4
Boiling Point 130° to 132°C (15mmHg)
Quantity 5g
Odor Almond-like
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.