Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Butylbenzoic acid, 99%

$72.01 - $742.84

Chemical Identifiers

Chemical Name or Material 4-n-Butylbenzoic acid
CAS 20651-71-2
Molecular Formula C11H14O2
Formula Weight 178.23
MDL Number MFCD00002571
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2389106

  SDS
alfa aesar™
B2389106
5g Each for $72.01

AAB2389114

  SDS
alfa aesar™
B2389114
25g Each for $239.58

AAB2389122

  SDS
alfa aesar™
B2389122
100g Each for $742.84
Description & Specifications

Specifications

Formula Weight 178.23
Melting Point 100° to 113°C
Health Hazard 1 Warning
Molecular Formula C11H14O2
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 5g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 4-n-Butylbenzoic acid
MDL Number MFCD00002571
TSCA TSCA
Assay 99%
CAS 20651-71-2
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.