Learn More
4-n-Butylbenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1657714
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-n-Butylbenzenesulfonyl chloride | |
13°C to 15°C | |
C10H13ClO2S | |
25 g | |
Moisture sensitive | |
OVFZELSNOHIDEF-UHFFFAOYSA-N | |
4-butylbenzenesulfonyl chloride | |
2737352 | |
97% |
54997-92-1 | |
126°C to 128°C (0.6 mmHg) | |
MFCD00173908 | |
UN3265 | |
4-butylbenzene-1-sulfonyl chloride, 4-n-butylbenzenesulfonyl chloride, 4-n-butyl benzenesulphonyl chloride, 4-butyl-benzenesulfonyl chloride, 4-n-butylphenylsulfonyl chloride, 4-n-butylbenzenesulphonyl chloride, benzenesulfonyl chloride, 4-butyl, 4-butylphenyl chlorosulfone, pubchem5484, 4-n-butylbenzenesulfonylchloride | |
CCCCC1=CC=C(C=C1)S(Cl)(=O)=O | |
232.72 | |
232.73 |
Chemical Identifiers
54997-92-1 | |
232.72 | |
OVFZELSNOHIDEF-UHFFFAOYSA-N | |
2737352 | |
CCCCC1=CC=C(C=C1)S(Cl)(=O)=O |
C10H13ClO2S | |
MFCD00173908 | |
4-butylbenzene-1-sulfonyl chloride, 4-n-butylbenzenesulfonyl chloride, 4-n-butyl benzenesulphonyl chloride, 4-butyl-benzenesulfonyl chloride, 4-n-butylphenylsulfonyl chloride, 4-n-butylbenzenesulphonyl chloride, benzenesulfonyl chloride, 4-butyl, 4-butylphenyl chlorosulfone, pubchem5484, 4-n-butylbenzenesulfonylchloride | |
4-butylbenzenesulfonyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: III
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only