Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Butylbenzenesulfonamide, 97+%

$29.09 - $100.68

Chemical Identifiers

Chemical Name or Material 4-n-Butylbenzenesulfonamide
CAS 1135-00-8
Molecular Formula C10H15NO2S
Formula Weight 213.29
MDL Number MFCD00465202
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2153403

 
alfa aesar™
B2153403
1g Each for $29.09

AAB2153406

 
alfa aesar™
B2153406
5g Each for $100.68
Description & Specifications

Specifications

Formula Weight 213.29
Chemical Name or Material 4-n-Butylbenzenesulfonamide
Assay ≥97%
Molecular Formula C10H15NO2S
Melting Point 93° to 95°C
MDL Number MFCD00465202
CAS 1135-00-8
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.