missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Butylbenzeneboronic acid, 98%

Manufacturer:  Alfa Aesar™ L1558406

 View more versions of this product

Catalog No. AAL1558406



Description

Description

Applications
Reactant for Suzuki-Miyaura cross-couplings, NHC-Iron-catalyzed aerobic oxidative aromatic esterification of aldehydes, Palladium-catalyzed oxidative Heck-type reactions.

Solubility
Soluble in methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

4-n-Butylbenzeneboronic acid
5g
C10H15BO2
6920034
UGZUUTHZEATQAM-UHFFFAOYSA-N
(4-butylphenyl)boronic acid
4100860
98%
103°C to 106°C
145240-28-4
MFCD02093926
4-butylbenzeneboronic acid
B(C1=CC=C(C=C1)CCCC)(O)O
178.038
178.04

Chemical Identifiers

145240-28-4
178.038
UGZUUTHZEATQAM-UHFFFAOYSA-N
4100860
B(C1=CC=C(C=C1)CCCC)(O)O
C10H15BO2
MFCD02093926
4-butylbenzeneboronic acid
(4-butylphenyl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents