Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Butylbenzaldehyde, 97%


Quantity 5g

Manufacturer: alfa aesar™  L1343806

 View more versions of this product

Catalog No. AAL1343806

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 162.23
Chemical Name or Material 4-n-Butylbenzaldehyde
MDL Number MFCD00040751
TSCA TSCA
Assay 97%
Molecular Formula C11H14O
Melting Point −43°C
CAS 1200-14-2
Beilstein 2208267
Quantity 5g
Boiling Point 96° to 98°C (5mmHg)

Specifications

Chemical Name or Material 4-n-Butylbenzaldehyde
CAS 1200-14-2
Molecular Formula C11H14O
Formula Weight 162.23
MDL Number MFCD00040751
Beilstein 2208267