Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Butylbenzaldehyde, 97%

$62.49 - $217.04

Chemical Identifiers

Chemical Name or Material 4-n-Butylbenzaldehyde
CAS 1200-14-2
Molecular Formula C11H14O
Formula Weight 162.23
MDL Number MFCD00040751
Beilstein 2208267
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1343803

 
alfa aesar™
L1343803
1g Each for $62.49

AAL1343806

 
alfa aesar™
L1343806
5g Each for $217.04
Description & Specifications

Specifications

Formula Weight 162.23
Chemical Name or Material 4-n-Butylbenzaldehyde
TSCA TSCA
Assay 97%
Molecular Formula C11H14O
MDL Number MFCD00040751
CAS 1200-14-2
Boiling Point 96° to 98°C (5mmHg)
Beilstein 2208267
Quantity 1g
Melting Point −43°C