Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Butylbenzaldehyde, 97%


Quantity 1g

Manufacturer: alfa aesar™  L1343803

 View more versions of this product

Catalog No. AAL1343803

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 162.23
Chemical Name or Material 4-n-Butylbenzaldehyde
MDL Number MFCD00040751
Assay 97%
CAS 1200-14-2
TSCA TSCA
Molecular Formula C11H14O
Beilstein 2208267
Quantity 1g
Boiling Point 96° to 98°C (5mmHg)
Melting Point −43°C

Specifications

Chemical Name or Material 4-n-Butylbenzaldehyde
CAS 1200-14-2
Molecular Formula C11H14O
Formula Weight 162.23
MDL Number MFCD00040751
Beilstein 2208267