Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Butylaniline, 97%

$37.85 - $261.68

Chemical Identifiers

Chemical Name or Material 4-n-Butylaniline
CAS 104-13-2
Molecular Formula C10H15N
Formula Weight 149.23
MDL Number MFCD00007925
Beilstein 1447268
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2523414

  SDS
alfa aesar™
B2523414
25g Each for $37.85

AAB2523422

  SDS
alfa aesar™
B2523422
100g Each for $82.97

AAB2523436

  SDS
alfa aesar™
B2523436
500g Each for $261.68
Description & Specifications

Specifications

Formula Weight 149.23
Chemical Name or Material 4-n-Butylaniline
TSCA TSCA
Assay 97%
Molecular Formula C10H15N
Melting Point −14°C
MDL Number MFCD00007925
CAS 104-13-2
Boiling Point 250°C
Beilstein 1447268
Quantity 25g
Flash Point 101°C (213°F)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number BW9470000
Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.