missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-n-Butylacetophenone, 97%

Manufacturer:  Alfa Aesar™ A1529614

 View more versions of this product

Catalog No. AAA1529614



Description

Description

Applications
4′-n-Butylacetophenone is used as an organic chemical synthesis intermediate.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, acids and bases.
Specifications

Specifications

4'-n-Butylacetophenone
101°C to 102°C (1.5mmHg)
37920-25-5
MFCD00017500
MQESVSITPLILCO-UHFFFAOYSA-N
1-(4-butylphenyl)ethanone
123470
97%
>110°C (230°F)
25g
C12H16O
4'-butylacetophenone, 1-4-butylphenyl ethanone, ethanone, 1-4-butylphenyl, p-butylacetophenone, 1-4-butylphenyl ethan-1-one, p-n-butylacetophenone, 4'-n-butylacetophenone, unii-6738cf9z8v, 1-acetyl-4-butylbenzene, 1-4-butyl-phenyl ethanone
CCCCC1=CC=C(C=C1)C(=O)C
176.259
176.26

Chemical Identifiers

37920-25-5
176.259
MQESVSITPLILCO-UHFFFAOYSA-N
123470
CCCCC1=CC=C(C=C1)C(=O)C
C12H16O
MFCD00017500
4'-butylacetophenone, 1-4-butylphenyl ethanone, ethanone, 1-4-butylphenyl, p-butylacetophenone, 1-4-butylphenyl ethan-1-one, p-n-butylacetophenone, 4'-n-butylacetophenone, unii-6738cf9z8v, 1-acetyl-4-butylbenzene, 1-4-butyl-phenyl ethanone
1-(4-butylphenyl)ethanone
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents