missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Butyl-4'-n-propylbicyclohexyl, 97%

$67.82 - $806.81

Chemical Identifiers

CAS 96624-52-1
Molecular Formula C19H36
Molecular Weight (g/mol) 264.497
MDL Number MFCD20260224
InChI Key QYGWJCMCLQAINT-UHFFFAOYSA-N
Synonym 1,1'-bicyclohexyl,4-butyl-4'-propyl-, trans,trans
PubChem CID 11086663
IUPAC Name 1-butyl-4-(4-propylcyclohexyl)cyclohexane
SMILES CCCCC1CCC(CC1)C2CCC(CC2)CCC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH55669MD View Documents Alfa Aesar™
H55669MD
250mg Each for $67.82
 
AAH5566903 View Documents Alfa Aesar™
H5566903
1g Each for $200.91
 
AAH5566906 View Documents Alfa Aesar™
H5566906
5g Each for $806.81
 
Specifications

Chemical Identifiers

96624-52-1
264.497
QYGWJCMCLQAINT-UHFFFAOYSA-N
11086663
CCCCC1CCC(CC1)C2CCC(CC2)CCC
C19H36
MFCD20260224
1,1'-bicyclohexyl,4-butyl-4'-propyl-, trans,trans
1-butyl-4-(4-propylcyclohexyl)cyclohexane

Specifications

83°C to 95°C
96624-52-1
MFCD20260224
QYGWJCMCLQAINT-UHFFFAOYSA-N
1-butyl-4-(4-propylcyclohexyl)cyclohexane
11086663
97%
250mg
C19H36
1,1'-bicyclohexyl,4-butyl-4'-propyl-, trans,trans
CCCCC1CCC(CC1)C2CCC(CC2)CCC
264.497
264.49
4-n-Butyl-4'-n-propylbicyclohexyl
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures