missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Butyl-4'-n-propylbicyclohexyl, 97%

Manufacturer:  Alfa Aesar™ H5566903

 View more versions of this product

Catalog No. AAH5566903



Specifications

Specifications

4-n-Butyl-4'-n-propylbicyclohexyl
1g
C19H36
1,1'-bicyclohexyl,4-butyl-4'-propyl-, trans,trans
CCCCC1CCC(CC1)C2CCC(CC2)CCC
264.497
264.49
83°C to 95°C
96624-52-1
MFCD20260224
QYGWJCMCLQAINT-UHFFFAOYSA-N
1-butyl-4-(4-propylcyclohexyl)cyclohexane
11086663
97%

Chemical Identifiers

96624-52-1
264.497
QYGWJCMCLQAINT-UHFFFAOYSA-N
11086663
CCCCC1CCC(CC1)C2CCC(CC2)CCC
C19H36
MFCD20260224
1,1'-bicyclohexyl,4-butyl-4'-propyl-, trans,trans
1-butyl-4-(4-propylcyclohexyl)cyclohexane
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents