Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Butyl-2-methylphenyl isothiocyanate, 95%

Chemical Identifiers

Chemical Name or Material 4-n-Butyl-2-methylphenyl isothiocyanate
CAS 175205-37-5
Molecular Formula C12H15NS
Formula Weight 205.32
MDL Number MFCD00276966
UN Number UN2922
Synonym p-n-Butyl-o-tolyl isothiocyanate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1898403

  SDS
alfa aesar™
A1898403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1898414

  SDS
alfa aesar™
A1898414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1898406

  SDS
alfa aesar™
A1898406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 205.32
Assay 95%
Molecular Formula C12H15NS
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 175205-37-5
UN Number UN2922
Synonym p-n-Butyl-o-tolyl isothiocyanate
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
Health Hazard 2 GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Quantity 1g
Chemical Name or Material 4-n-Butyl-2-methylphenyl isothiocyanate
MDL Number MFCD00276966
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.