Learn More
4-n-Butoxyphenol, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1207614
Description
It is an intermediate of liquid crystals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an intermediate of liquid crystals.
Solubility
Soluble in acetone and ethanol.
Notes
Store in cool place. Keep away from oxidizing agents.
Specifications
4-n-Butoxyphenol | |
63°C to 66°C | |
MFCD00002336 | |
phenol, 4-butoxy, p-butoxyphenol, 4-n-butoxyphenol, phenol, p-butoxy, hydroquinone monobutyl ether, unii-8188dbs68j, 4-normal-butoxyphenol, 4-butoxy-pheno, 4-butoxy-phenol, 4-n-butyloxyphenol | |
MBGGFXOXUIDRJD-UHFFFAOYSA-N | |
4-butoxyphenol | |
31233 | |
166.22 |
122-94-1 | |
C10H14O2 | |
25 g | |
Soluble in acetone and ethanol. | |
CCCCOC1=CC=C(O)C=C1 | |
166.22 | |
CHEBI:34392 | |
98% |
Chemical Identifiers
122-94-1 | |
166.22 | |
MBGGFXOXUIDRJD-UHFFFAOYSA-N | |
31233 | |
4-butoxyphenol |
C10H14O2 | |
MFCD00002336 | |
phenol, 4-butoxy, p-butoxyphenol, 4-n-butoxyphenol, phenol, p-butoxy, hydroquinone monobutyl ether, unii-8188dbs68j, 4-normal-butoxyphenol, 4-butoxy-pheno, 4-butoxy-phenol, 4-n-butyloxyphenol | |
CHEBI:34392 | |
CCCCOC1=CC=C(O)C=C1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 204-583-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only