Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(n-Butoxycarbonyl)phenyl isocyanate, 99%

Chemical Identifiers

Chemical Name or Material 4-(n-Butoxycarbonyl)phenyl isocyanate
CAS 102561-47-7
Molecular Formula C12H13NO3
Formula Weight 219.24
MDL Number MFCD00037043
UN Number UN2206
Beilstein 7640773
Synonym n-Butyl 4-isocyanatobenzoate; 4-(Carbobutoxy)phenyl isocyanate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1060603

  SDS
alfa aesar™
L1060603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1060606

  SDS
alfa aesar™
L1060606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 219.24
CAS 102561-47-7
MDL Number MFCD00037043
Health Hazard 2 GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Danger
Chemical Name or Material 4-(n-Butoxycarbonyl)phenyl isocyanate
UN Number UN2206
Synonym n-Butyl 4-isocyanatobenzoate; 4-(Carbobutoxy)phenyl isocyanate
Health Hazard 3 GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Molecular Formula C12H13NO3
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
Beilstein 7640773
Assay 99%
Quantity 1g
Recommended Storage 2° to 8°C
Flash Point >110°C (230°F)
Boiling Point 295°C
Safety and Handling

Danger

GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C