missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(n-Butoxycarbonyl)phenyl isocyanate, 99%

Manufacturer:  Alfa Aesar™ L1060603

 View more versions of this product

Catalog No. AAL1060603



Specifications

Specifications

4-(n-Butoxycarbonyl)phenyl isocyanate
295°C
102561-47-7
MFCD00037043
7640773
PVCGQSQNUXNTNA-UHFFFAOYSA-N
butyl 4-isocyanatobenzoate
4112112
99%
>110°C (230°F)
1g
C12H13NO3
UN2206
4-n-butoxycarbonyl phenyl isocyanate, acmc-20e5xd, 4-carbobutoxy phenyl isocyanate, butyl 4-isocyanatobenzoate, 4-n-butoxycarbonyl phenylisocyanate, 4-carbobutoxyphenyl isocyanate, 4-n-carbobutoxyphenyl isocyanate, benzoic acid,4-isocyanato-, butyl ester, benzoic acid, 4-isocyanato-, butyl ester 9ci, 4-n-butoxycarbonyl phenyl isocyanate; 4-carbobutoxy phenyl isocyanate
CCCCOC(=O)C1=CC=C(C=C1)N=C=O
219.24
219.24

Chemical Identifiers

102561-47-7
219.24
PVCGQSQNUXNTNA-UHFFFAOYSA-N
4112112
CCCCOC(=O)C1=CC=C(C=C1)N=C=O
C12H13NO3
MFCD00037043
4-n-butoxycarbonyl phenyl isocyanate, acmc-20e5xd, 4-carbobutoxy phenyl isocyanate, butyl 4-isocyanatobenzoate, 4-n-butoxycarbonyl phenylisocyanate, 4-carbobutoxyphenyl isocyanate, 4-n-carbobutoxyphenyl isocyanate, benzoic acid,4-isocyanato-, butyl ester, benzoic acid, 4-isocyanato-, butyl ester 9ci, 4-n-butoxycarbonyl phenyl isocyanate; 4-carbobutoxy phenyl isocyanate
butyl 4-isocyanatobenzoate
Safety and Handling

Safety and Handling

GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.

Recommended Storage : 2° to 8°C

Documents