Learn More
4-(n-Butoxycarbonyl)phenyl isocyanate, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals L1060606
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-(n-Butoxycarbonyl)phenyl isocyanate | |
1.105 | |
>110°C (230°F) | |
1.5275 | |
5 g | |
7640773 | |
4-n-butoxycarbonyl phenyl isocyanate, acmc-20e5xd, 4-carbobutoxy phenyl isocyanate, butyl 4-isocyanatobenzoate, 4-n-butoxycarbonyl phenylisocyanate, 4-carbobutoxyphenyl isocyanate, 4-n-carbobutoxyphenyl isocyanate, benzoic acid,4-isocyanato-, butyl ester, benzoic acid, 4-isocyanato-, butyl ester 9ci, 4-n-butoxycarbonyl phenyl isocyanate; 4-carbobutoxy phenyl isocyanate | |
CCCCOC(=O)C1=CC=C(C=C1)N=C=O | |
219.24 | |
219.24 |
102561-47-7 | |
295°C | |
C12H13NO3 | |
MFCD00037043 | |
UN2206 | |
Moisture sensitive | |
PVCGQSQNUXNTNA-UHFFFAOYSA-N | |
butyl 4-isocyanatobenzoate | |
4112112 | |
99% |
Chemical Identifiers
102561-47-7 | |
219.24 | |
PVCGQSQNUXNTNA-UHFFFAOYSA-N | |
4112112 | |
CCCCOC(=O)C1=CC=C(C=C1)N=C=O |
C12H13NO3 | |
MFCD00037043 | |
4-n-butoxycarbonyl phenyl isocyanate, acmc-20e5xd, 4-carbobutoxy phenyl isocyanate, butyl 4-isocyanatobenzoate, 4-n-butoxycarbonyl phenylisocyanate, 4-carbobutoxyphenyl isocyanate, 4-n-carbobutoxyphenyl isocyanate, benzoic acid,4-isocyanato-, butyl ester, benzoic acid, 4-isocyanato-, butyl ester 9ci, 4-n-butoxycarbonyl phenyl isocyanate; 4-carbobutoxy phenyl isocyanate | |
butyl 4-isocyanatobenzoate |
Safety and Handling
GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P311-P330-P333+P313-P362-P501c
H302-H315-H317-H319-H331-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
TSCA : No
Recommended Storage : Keep cold