missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Butoxybenzaldehyde, 98%

$55.04 - $55.04

Chemical Identifiers

CAS 5736-88-9
Molecular Formula C11H14O2
Formula Weight 178.23
MDL Number MFCD00003389
Chemical Name or Material 4-n-Butoxybenzaldehyde
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1524309 SDS Alfa Aesar™
A1524309
10g Each for $55.04
AAA1524318 SDS Alfa Aesar™
A1524318
50g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

5736-88-9
178.23
4-n-Butoxybenzaldehyde
C11H14O2
MFCD00003389

Specifications

5736-88-9
178.23
284° to 286°C
>110°C (230°F)
98%
C11H14O2
MFCD00003389
10g
4-n-Butoxybenzaldehyde

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA