Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-n-Butoxyacetophenone, 98%

$100.20 - $363.14

Chemical Identifiers

Chemical Name or Material 4'-n-Butoxyacetophenone
CAS 5736-89-0
Molecular Formula C12H16O2
Formula Weight 192.26
MDL Number MFCD00027200
Beilstein 637894
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0846806

  SDS
alfa aesar™
L0846806
5g Each for $100.20

AAL0846814

  SDS
alfa aesar™
L0846814
25g Each for $363.14
Description & Specifications

Specifications

Formula Weight 192.26
Chemical Name or Material 4'-n-Butoxyacetophenone
MDL Number MFCD00027200
Assay 98%
Molecular Formula C12H16O2
TSCA TSCA
CAS 5736-89-0
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P301+P312-P321-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Specific treatment (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Boiling Point 172° to 173°C (18mmHg)
Melting Point 25° to 27°C
Beilstein 637894
Odor Sweet
Flash Point >110°C (230°F)
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P301+P312-P321-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Specific treatment (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.