Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(n-Butoxy)phenylacetic acid, 98+%

$44.15 - $44.15

Chemical Identifiers

Chemical Name or Material 4-(n-Butoxy)phenylacetic acid
CAS 4547-57-3
Molecular Formula C12H16O3
Formula Weight 208.26
MDL Number MFCD00016832
Beilstein 1105534
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0823103

  SDS
alfa aesar™
L0823103
1g Each for $44.15
Description & Specifications

Specifications

Formula Weight 208.26
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay ≥98%
Chemical Name or Material 4-(n-Butoxy)phenylacetic acid
Health Hazard 1 Warning
MDL Number MFCD00016832
Beilstein 1105534
Molecular Formula C12H16O3
Melting Point 87° to 89°C
RTECS Number AF6945000
Quantity 1g
CAS 4547-57-3
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.