missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-N-BOC-Amino-4-carboxytetrahydropyran, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC360460020

 View more versions of this product

Catalog No. AC360460020

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥94% (Acid)
245.27
White
100.0
4-N-BOC-Amino-4-carboxytetrahydropyran
C11H19NO5
95%
Powder
2g
94.0
Authentic
172843-97-9
MFCD02683136
95%

Chemical Identifiers

172843-97-9
100.0
C11H19NO5
MFCD02683136
≥94% (Acid)
94.0
95%
245.27
4-N-BOC-Amino-4-carboxytetrahydropyran, 95%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.