Please login to your online account to display your discounted pricing

4-N-BOC-Amino-4-carboxytetrahydropyran, 95%, ACROS Organics™


Quantity 500mg

Manufacturer: thermo fisher scientific  36046-5000

 View more versions of this product

Catalog No. AC360465000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94% (Acid)
Physical Form Powder
Formula Weight 245.27
Quantity 500mg
Color White
CAS Max % 100.0
CAS Min % 94.0
Assay Percent Range 95%
Chemical Name or Material 4-N-BOC-Amino-4-carboxytetrahydropyran
CAS 172843-97-9
Name Note 95%
Molecular Formula C11H19NO5
MDL Number MFCD02683136
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-N-BOC-Amino-4-carboxytetrahydropyran
Name Note 95%
CAS 172843-97-9
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94% (Acid)
Assay Percent Range 95%
Molecular Formula C11H19NO5
Formula Weight 245.27
MDL Number MFCD02683136
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents