Learn More
4-Morpholinylsulfur trifluoride, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1975106
Description
4-Morpholinylsulfur trifluoride is incorporated in the fluoronation of trihexanols. It is used as a nucleophilic fluorinating agent with greater thermal stability. It is also used in the preparation of proline analogs and in the preparation of difluoropiperidines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Morpholinylsulfur trifluoride is incorporated in the fluoronation of trihexanols. It is used as a nucleophilic fluorinating agent with greater thermal stability. It is also used in the preparation of proline analogs and in the preparation of difluoropiperidines.
Solubility
Reacts with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with bases and oxidizing and bases. Store under dry inert gas. It is sensitive to moisture.
Specifications
4-Morpholinylsulfur trifluoride | |
1.436 | |
76°C (168°F) | |
1.4545 | |
5 g | |
606224 | |
morpholinosulfur trifluoride, morpholinosulfurtrifluoride, morph-dast, 4-trifluoro-$l^ 4-sulfanyl morpholine, 4-trifluoro-??-sulfanyl morpholine, morpholinosulphur trifluoride, morpho-dast, acmc-209krh, trifluoro morpholin-4-yl, morpholinotrifluorosulfur iv | |
UFXIRMVZNARBDL-UHFFFAOYSA-N | |
trifluoro(morpholin-4-yl)-$l^{4}-sulfane | |
2733258 | |
95% |
51010-74-3 | |
41°C to 42°C (0.5 mmHg) | |
C4H8F3NOS | |
MFCD00037057 | |
UN3265 | |
Moisture sensitive; May form a precipitate | |
Reacts with water. | |
C1COCCN1S(F)(F)F | |
175.169 | |
175.17 |
Chemical Identifiers
51010-74-3 | |
175.169 | |
UFXIRMVZNARBDL-UHFFFAOYSA-N | |
2733258 | |
C1COCCN1S(F)(F)F |
C4H8F3NOS | |
MFCD00037057 | |
morpholinosulfur trifluoride, morpholinosulfurtrifluoride, morph-dast, 4-trifluoro-$l^ 4-sulfanyl morpholine, 4-trifluoro-??-sulfanyl morpholine, morpholinosulphur trifluoride, morpho-dast, acmc-209krh, trifluoro morpholin-4-yl, morpholinotrifluorosulfur iv | |
trifluoro(morpholin-4-yl)-$l^{4}-sulfane |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only