Please login to your online account to display your discounted pricing

4-(Morpholinosulfonyl)aniline, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 4-(Morpholinosulfonyl)aniline
CAS 21626-70-0
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H14N2O3S
Formula Weight 242.3
MDL Number MFCD00277213
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

TL00826EA

 
fisher scientific company
TL00826EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 242.3
Quantity 10g
Assay 97%
CAS Min % 97.0
Molecular Formula C10H14N2O3S
CAS 21626-70-0
Assay Percent Range 97%
Chemical Name or Material 4-(Morpholinosulfonyl)aniline
MDL Number MFCD00277213
Packaging Amber Glass Bottle