Please login to your online account to display your discounted pricing

4-(Morpholinosulfonyl)aniline, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  TL00826EA

 View more versions of this product

Catalog No. TL00826EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 242.3
Quantity 10g
Assay 97%
CAS Min % 97.0
Molecular Formula C10H14N2O3S
CAS 21626-70-0
Assay Percent Range 97%
Chemical Name or Material 4-(Morpholinosulfonyl)aniline
MDL Number MFCD00277213
Packaging Amber Glass Bottle

Specifications

Chemical Name or Material 4-(Morpholinosulfonyl)aniline
CAS 21626-70-0
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H14N2O3S
Formula Weight 242.3
MDL Number MFCD00277213