Please login to your online account to display your discounted pricing

4-Morpholinophenylboronic acid, 97%, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC17412DE

 View more versions of this product

Catalog No. CC17412DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 207.04
Quantity 5g
Assay 97%
Packaging Amber Glass Bottle
Assay Percent Range 97%
CAS 186498-02-2
Chemical Name or Material 4-Morpholinophenylboronic acid
Molecular Formula C10H14BNO3

Specifications

Chemical Name or Material 4-Morpholinophenylboronic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H14BNO3
Formula Weight 207.04