missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Morpholinophenyl isothiocyanate, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC17407DE
Specifications
4-Morpholinophenyl isothiocyanate | |
97% | |
C11H12N2OS | |
4-morpholinophenyl isothiocyanate, 4-4-isothiocyanatophenyl morpholine, 4-morpholinophenylisothiocyanate, morpholine, 4-4-isothiocyanatophenyl, 4-4-isothiocyanatophenyl morpholin, 4-morpholin-4-ylbenzenisothiocyanate, 4-4-morpholinyl phenyl isothiocyanate, 4-4-isothiocyanato-phenyl-morpholine, morpholine,4-4-isothiocyanatophenyl, 4-morpholin-4-yl phenyl isothiocyanate | |
C1COCCN1C2=CC=C(C=C2)N=C=S | |
220.29 | |
220.29 |
51317-66-9 | |
Amber Glass Bottle | |
5g | |
AXUXRZZYZBZQAR-UHFFFAOYSA-N | |
4-(4-isothiocyanatophenyl)morpholine | |
224862 | |
97% |
Chemical Identifiers
51317-66-9 | |
220.29 | |
4-morpholinophenyl isothiocyanate, 4-4-isothiocyanatophenyl morpholine, 4-morpholinophenylisothiocyanate, morpholine, 4-4-isothiocyanatophenyl, 4-4-isothiocyanatophenyl morpholin, 4-morpholin-4-ylbenzenisothiocyanate, 4-4-morpholinyl phenyl isothiocyanate, 4-4-isothiocyanato-phenyl-morpholine, morpholine,4-4-isothiocyanatophenyl, 4-morpholin-4-yl phenyl isothiocyanate | |
4-(4-isothiocyanatophenyl)morpholine |
C11H12N2OS | |
AXUXRZZYZBZQAR-UHFFFAOYSA-N | |
224862 | |
C1COCCN1C2=CC=C(C=C2)N=C=S |