missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Morpholinomethyl)benzaldehyde, 97%, Maybridge

$82.77 - $1,195.91

Chemical Identifiers

CAS 82413-63-6
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C12H15NO2
Formula Weight 205.26
MDL Number MFCD05865107
Synonym 4-(morpholinomethyl)benzaldehyde (IUPAC)
Chemical Name or Material 4-(Morpholinomethyl)benzaldehyde
Assay 95%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC29304EA Fisher Scientific Company
CC29304EA
Amber Glass Bottle 10g Each for $1,195.91
CC29304DA SDS Fisher Scientific Company
CC29304DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC29304CB SDS Fisher Scientific Company
CC29304CB
Amber Glass Bottle 250mg Each for $82.77
Specifications Description & Specifications

Chemical Identifiers

82413-63-6
100.0
C12H15NO2
MFCD05865107
4-(Morpholinomethyl)benzaldehyde
94.5
95%
205.26
4-(morpholinomethyl)benzaldehyde (IUPAC)
95%

Specifications

82413-63-6
100.0
C12H15NO2
MFCD05865107
4-(morpholinomethyl)benzaldehyde (IUPAC)
4-(Morpholinomethyl)benzaldehyde
94.5
95%
205.26
Amber Glass Bottle
10g
95%

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger