Please login to your online account to display your discounted pricing

4-(Morpholinomethyl)aniline, 95+%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC29314CB

 View more versions of this product

Catalog No. CC29314CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 192.26
Quantity 250mg
Assay 95%
Chemical Name or Material 4-(Morpholinomethyl)aniline
CAS 51013-67-3
Packaging Amber Glass Bottle
Assay Percent Range 95%
Molecular Formula C11H16N2O

Specifications

Chemical Name or Material 4-(Morpholinomethyl)aniline
Assay 95%
Assay Percent Range 95%
Molecular Formula C11H16N2O
Formula Weight 192.26