missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Morpholinobenzoic acid, 97%, Maybridge

$13.82 - $13.82

Chemical Identifiers

CAS 7470-38-4
Molecular Formula C11H13NO3
Molecular Weight (g/mol) 207.229
MDL Number MFCD00588379
InChI Key XVAJKPNTGSKZSQ-UHFFFAOYSA-N
Synonym 4-4-morpholinyl benzoic acid
PubChem CID 345278
IUPAC Name 4-morpholin-4-ylbenzoic acid
SMILES C1COCCN1C2=CC=C(C=C2)C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC17401CB SDS View Documents Fisher Scientific Company
CC17401CB
250mg Amber Glass Bottle Each for $13.82
 
Specifications

Chemical Identifiers

7470-38-4
207.229
XVAJKPNTGSKZSQ-UHFFFAOYSA-N
345278
C1COCCN1C2=CC=C(C=C2)C(=O)O
C11H13NO3
MFCD00588379
4-4-morpholinyl benzoic acid
4-morpholin-4-ylbenzoic acid

Specifications

Amber Glass Bottle
7470-38-4
100.0
C11H13NO3
4-4-morpholinyl benzoic acid
C1COCCN1C2=CC=C(C=C2)C(=O)O
207.229
207.23
4-Morpholinobenzoic acid
250mg
96.5
97%
MFCD00588379
XVAJKPNTGSKZSQ-UHFFFAOYSA-N
4-morpholin-4-ylbenzoic acid
345278
97%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.