missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Morpholinoaniline, 98+%, ACROS Organics™

$94.45 - $94.45

Chemical Identifiers

CAS 2524-67-6
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Molecular Formula C10H14N2O
Formula Weight 178.23
MDL Number MFCD00006169
Chemical Name or Material 4-Morpholinoaniline
Assay ≥98.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC188650250 SDS Thermo Fisher Scientific
AC188650250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC188650050 SDS Thermo Fisher Scientific
188650050
Glass bottle 5g Each for $94.45
Specifications Description & Specifications

Chemical Identifiers

2524-67-6
100.0
C10H14N2O
MFCD00006169
≥98.0%
98.0
98+%
178.23
4-Morpholinoaniline

Specifications

Crystalline Powder
25g
128° to 132°C
98.0
2524-67-6
MFCD00006169
Authentic
≥98.0%
178.23
Purple to brown
100.0
C10H14N2O
98+%
Glass bottle
4-Morpholinoaniline

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning