Learn More
4-Morpholinoaniline, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 188650050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Morpholinoaniline | |
98.0 | |
130°C to 131°C | |
Authentic | |
Glass bottle | |
MFCD00006169 | |
4-morpholinoaniline, 4-morpholin-4-yl-phenylamine, 4-morpholin-4-yl aniline, n-4-aminophenyl morpholine, p-morpholinoaniline, 4-morpholinobenzenamine, 4-4-morpholino aniline, benzenamine, 4-4-morpholinyl, 4-morpholinyl aniline, 4-4-aminophenyl-morpholine | |
C1COCCN1C2=CC=C(C=C2)N | |
178.23 | |
178.23 | |
Crystalline Powder |
2524-67-6 | |
100.0 | |
Purple to Brown | |
98+% | |
C10H14N2O | |
5 g | |
PHNDZBFLOPIMSM-UHFFFAOYSA-N | |
4-morpholin-4-ylaniline | |
75655 | |
98+% |
Chemical Identifiers
2524-67-6 | |
178.23 | |
PHNDZBFLOPIMSM-UHFFFAOYSA-N | |
75655 | |
C1COCCN1C2=CC=C(C=C2)N |
C10H14N2O | |
MFCD00006169 | |
4-morpholinoaniline, 4-morpholin-4-yl-phenylamine, 4-morpholin-4-yl aniline, n-4-aminophenyl morpholine, p-morpholinoaniline, 4-morpholinobenzenamine, 4-4-morpholino aniline, benzenamine, 4-4-morpholinyl, 4-morpholinyl aniline, 4-4-aminophenyl-morpholine | |
4-morpholin-4-ylaniline |
Safety and Handling
GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 219-760-1
RUO – Research Use Only