missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Morpholinoaniline, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals BTB01373EB

 View more versions of this product

Catalog No. BTB01373EB



Description

Description

Specifications

Specifications

4-Morpholinoaniline
96.5
97%
C10H14N2O
25 g
PHNDZBFLOPIMSM-UHFFFAOYSA-N
4-morpholin-4-ylaniline
75655
97%
2524-67-6
100.0
Amber Glass Bottle
MFCD00006169
4-morpholinoaniline, 4-morpholin-4-yl-phenylamine, 4-morpholin-4-yl aniline, n-4-aminophenyl morpholine, p-morpholinoaniline, 4-morpholinobenzenamine, 4-4-morpholino aniline, benzenamine, 4-4-morpholinyl, 4-morpholinyl aniline, 4-4-aminophenyl-morpholine
C1COCCN1C2=CC=C(C=C2)N
178.235
178.23

Chemical Identifiers

2524-67-6
178.235
PHNDZBFLOPIMSM-UHFFFAOYSA-N
75655
C1COCCN1C2=CC=C(C=C2)N
C10H14N2O
MFCD00006169
4-morpholinoaniline, 4-morpholin-4-yl-phenylamine, 4-morpholin-4-yl aniline, n-4-aminophenyl morpholine, p-morpholinoaniline, 4-morpholinobenzenamine, 4-4-morpholino aniline, benzenamine, 4-4-morpholinyl, 4-morpholinyl aniline, 4-4-aminophenyl-morpholine
4-morpholin-4-ylaniline
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions