missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Morpholineacetic acid hydrochloride, 95%

Manufacturer:  Alfa Aesar™ H61188MD

 View more versions of this product

Catalog No. AAH61188MD



Description

Description

Applications
4-Morpholineacetic acid hydrochloride is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

4-Morpholineacetic acid hydrochloride
250mg
C6H12ClNO3
3701710
Insoluble in water.
C1COCCN1CC(=O)O.Cl
181.616
181.62
95%
White
89531-58-8
MFCD03540888
2-morpholin-4-yl acetic acid hydrochloride, 2-morpholinoacetic acid hydrochloride, 4-morpholinacetic acid hcl, 4-morpholineacetic acid hydrochloride, 4-morpholineacetic acid, hydrochloride, 4-morpholineaceticacid, hydrochloride, 4-morpholineaceticacid, hydrochloride 1:1, 4-morpholinylacetic acid hydrochloride, morpholin-4-yl-acetic acid hydrochloride, morpholin-4-ylacetic acid hydrochloride
ICGYBJNONRDZOP-UHFFFAOYSA-N
2-morpholin-4-ylacetic acid;hydrochloride
17750946
Powder

Chemical Identifiers

89531-58-8
181.616
ICGYBJNONRDZOP-UHFFFAOYSA-N
17750946
C1COCCN1CC(=O)O.Cl
C6H12ClNO3
MFCD03540888
2-morpholin-4-yl acetic acid hydrochloride, 2-morpholinoacetic acid hydrochloride, 4-morpholinacetic acid hcl, 4-morpholineacetic acid hydrochloride, 4-morpholineacetic acid, hydrochloride, 4-morpholineaceticacid, hydrochloride, 4-morpholineaceticacid, hydrochloride 1:1, 4-morpholinylacetic acid hydrochloride, morpholin-4-yl-acetic acid hydrochloride, morpholin-4-ylacetic acid hydrochloride
2-morpholin-4-ylacetic acid;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents