missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(4-Morpholin-4-yl-phenyl)-methanol, 95+%, Maybridge

Manufacturer:  Thermo Scientific CC17409DA

 View more versions of this product

Catalog No. CC17409DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

(4-Morpholin-4-yl-phenyl)-methanol
1g
95%
4-morpholin-4-yl-phenyl methanol, 4-morpholinobenzyl alcohol, 4-morpholin-4-yl phenyl methanol, 4-morpholinophenyl methanol, 4-morpholin-4-yl-phenyl-methanol, 4-morpholin-4-ylphenyl methanol, 4-morpholin-4-yl benzyl alcohol, 4-morpholin-4-ylphenyl methan-1-ol, 4-morpholinobenzylalcohol, 4-4-morpholinyl benzenemethanol
C1COCCN1C2=CC=C(C=C2)CO
193.246
193.24
Amber Glass Bottle
280556-71-0
C11H15NO2
KUAHZNZWSIDSTH-UHFFFAOYSA-N
(4-morpholin-4-ylphenyl)methanol
2776456
95%

Chemical Identifiers

280556-71-0
193.246
4-morpholin-4-yl-phenyl methanol, 4-morpholinobenzyl alcohol, 4-morpholin-4-yl phenyl methanol, 4-morpholinophenyl methanol, 4-morpholin-4-yl-phenyl-methanol, 4-morpholin-4-ylphenyl methanol, 4-morpholin-4-yl benzyl alcohol, 4-morpholin-4-ylphenyl methan-1-ol, 4-morpholinobenzylalcohol, 4-4-morpholinyl benzenemethanol
(4-morpholin-4-ylphenyl)methanol
C11H15NO2
KUAHZNZWSIDSTH-UHFFFAOYSA-N
2776456
C1COCCN1C2=CC=C(C=C2)CO
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS Documents