missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Methylvaleric Acid 97.0+%, TCI America™
Supplier: TCI America M075025ML
Specifications
4-Methylvaleric Acid | |
Yellow | |
C6H12O2 | |
25 mL | |
4-methylvaleric acid, isocaproic acid, isohexanoic acid, pentanoic acid, 4-methyl, isobutylacetic acid, 4-methyl-pentanoic acid, valeric acid, 4-methyl, 4-methyl valeric acid, isocaproate, 4,4-dimethylbutanoic acid | |
CC(C)CCC(=O)O | |
116.16 | |
CHEBI:74903 | |
≥97.0% (GC) |
646-07-1 | |
201°C | |
MFCD00002803 | |
2810 | |
FGKJLKRYENPLQH-UHFFFAOYSA-N | |
4-methylpentanoic acid | |
12587 | |
116.16 | |
Liquid |
Chemical Identifiers
646-07-1 | |
116.16 | |
FGKJLKRYENPLQH-UHFFFAOYSA-N | |
12587 | |
4-methylpentanoic acid |
C6H12O2 | |
MFCD00002803 | |
4-methylvaleric acid, isocaproic acid, isohexanoic acid, pentanoic acid, 4-methyl, isobutylacetic acid, 4-methyl-pentanoic acid, valeric acid, 4-methyl, 4-methyl valeric acid, isocaproate, 4,4-dimethylbutanoic acid | |
CHEBI:74903 | |
CC(C)CCC(=O)O |
Safety and Handling
EINECSNumber : (2)-0608
RTECSNumber : NR2975000
TSCA : Yes