missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Methylvaleric acid, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 255510250
Specifications
4-Methylvaleric acid | |
97°C | |
Glass bottle | |
0.922 | |
Colorless to Yellow | |
25mL | |
646-07-1 | |
100.0 | |
C6H12O2 | |
MFCD00002803 | |
Solubility in water: insoluble | |
CC(C)CCC(=O)O | |
116.15 | |
CHEBI:74903 | |
Liquid |
0.9220g/mL | |
Authentic | |
1.4136 to 1.4156 | |
200.0°C | |
-35.0°C | |
99% | |
98.5 | |
98.5% min. (GC) | |
(CH3)2CHCH2CH2COOH | |
4,4-dimethylbutanoic acid, 4-methyl valeric acid, 4-methyl-pentanoic acid, 4-methylvaleric acid, isobutylacetic acid, isocaproate, isocaproic acid, isohexanoic acid, pentanoic acid, 4-methyl, valeric acid, 4-methyl | |
FGKJLKRYENPLQH-UHFFFAOYSA-N | |
4-methylpentanoic acid | |
12587 | |
116.15 | |
99% |
Chemical Identifiers
646-07-1 | |
116.15 | |
FGKJLKRYENPLQH-UHFFFAOYSA-N | |
12587 | |
4-methylpentanoic acid |
C6H12O2 | |
MFCD00002803 | |
4,4-dimethylbutanoic acid, 4-methyl valeric acid, 4-methyl-pentanoic acid, 4-methylvaleric acid, isobutylacetic acid, isocaproate, isocaproic acid, isohexanoic acid, pentanoic acid, 4-methyl, valeric acid, 4-methyl | |
CHEBI:74903 | |
CC(C)CCC(=O)O |
Safety and Handling
GHS H Statement
Toxic in contact with skin.
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
EINECSNumber : 211-464-0
RTECSNumber : NR2975000
TSCA : TSCA