missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Methylumbelliferyl phosphate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals J66091MD
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Methylumbelliferyl phosphate | |
214°C to 216°C | |
C10H9O6P | |
250 mg | |
4-methylumbelliferyl phosphate, 4-methyl-2-oxo-2h-chromen-7-yl dihydrogen phosphate, 4-mup, 2h-1-benzopyran-2-one, 4-methyl-7-phosphonooxy, phosphoric acid 4-methylumbelliferyl ester, 4-methylumbelliferylphosphate, free acid, 4-methyl-2-oxochromen-7-yl oxyphosphonic acid, 5,5-dinitrobarbutyric acid, 4-methylumbelliferone phosphate, 4-methylumbelliferyl phosphate, free acid | |
BCHIXGBGRHLSBE-UHFFFAOYSA-N | |
(4-methyl-2-oxochromen-7-yl) dihydrogen phosphate | |
65118 | |
99% |
3368-04-5 | |
White | |
MFCD00016969 | |
1687229 | |
Soluble in water | |
CC1=CC(=O)OC2=C1C=CC(=C2)OP(=O)(O)O | |
256.15 | |
256.15 | |
Powder |
Chemical Identifiers
3368-04-5 | |
MFCD00016969 |
Safety and Handling
EINECSNumber : 222-137-7
TSCA : Yes
Recommended Storage : Store at -20°C
RUO – Research Use Only