missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Methylumbelliferyl-β-D-glucopyranoside, 99%, Thermo Scientific Chemicals
$1189.33 - $1189.33
Chemical Identifiers
CAS | 18997-57-4 |
---|---|
Molecular Formula | C16H18O8 |
Molecular Weight (g/mol) | 338.31 |
MDL Number | MFCD00063694 |
InChI Key | YUDPTGPSBJVHCN-YMILTQATSA-N |
Synonym | 4-methylumbelliferyl, a-d-glucoside, glc1-b-4mu, 7-, a-d-glucopyranosyloxy-4-methyl-2h-1-benzopyran-2-one, 4-methylumbelliferyl glucopyranoside, 4-methyl-7-2s,4s,5s-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy chromen-2-one |
PubChem CID | 2733779 |
IUPAC Name | 4-methyl-7-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-2-one |
SMILES | CC1=CC(=O)OC2=C1C=CC(=C2)OC3C(C(C(C(O3)CO)O)O)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC337020025
|
Thermo Scientific Chemicals
337020025 |
2.5 g | Glass bottle |
Each for $1,189.33
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
18997-57-4 | |
338.31 | |
YUDPTGPSBJVHCN-YMILTQATSA-N | |
2733779 | |
CC1=CC(=O)OC2=C1C=CC(=C2)OC3C(C(C(C(O3)CO)O)O)O |
C16H18O8 | |
MFCD00063694 | |
4-methylumbelliferyl, a-d-glucoside, glc1-b-4mu, 7-, a-d-glucopyranosyloxy-4-methyl-2h-1-benzopyran-2-one, 4-methylumbelliferyl glucopyranoside, 4-methyl-7-2s,4s,5s-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy chromen-2-one | |
4-methyl-7-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-2-one |
Specifications
18997-57-4 | |
100.0 | |
White | |
99% | |
C16H18O8 | |
2.5 g | |
−99.00 (20.00°C c=0.3,water) | |
4-methylumbelliferyl, a-d-glucoside, glc1-b-4mu, 7-, a-d-glucopyranosyloxy-4-methyl-2h-1-benzopyran-2-one, 4-methylumbelliferyl glucopyranoside, 4-methyl-7-2s,4s,5s-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy chromen-2-one | |
YUDPTGPSBJVHCN-YMILTQATSA-N | |
4-methyl-7-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-2-one | |
2733779 | |
99% | |
4-Methylumbelliferyl-β-D-glucopyranoside, 99% |
98.5 | |
210.0°C to 212.0°C | |
Authentic | |
Glass bottle | |
MFCD00063694 | |
18, V,1, 443 | |
-99 | |
Solubility in water: moderately soluble. Other solubilities: soluble in ethanol,2-propanol,acetone,pyridine | |
CC1=CC(=O)OC2=C1C=CC(=C2)OC3C(C(C(C(O3)CO)O)O)O | |
338.31 | |
338.31 | |
Powder |
Safety and Handling
MOISTURE SENSITIVE
EINECSNumber : 242-736-7
RUO – Research Use Only